Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spk-1
Entrez Gene ID
176328
Gene Name
protein-coding
Gene Aliases
CELE_B0464.5
Chromosome Location
Chr.III: 9456608 - 9463007 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025962.3 NP_001021133.1 3 - 4 323 85
NM_001025963.6 NP_001021134.1 3 - 4 329 85
NM_066679.5 NP_499080.3 2 - 3 1615 85
GenBank mRNA AF241656.1 - 3 - 4 376 85

Gene Symbol
B0464.9
Entrez Gene ID
181999
Gene Name
Probable protein phosphatase methylesterase 1
Gene Aliases
CELE_B0464.9
Chromosome Location
Chr.III: 9456604 - 9467180 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066683.5 NP_499084.1 3 - 4 333 85

View Full Product Details