Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spk-1
Entrez Gene ID
176328
Gene Name
protein-coding
Gene Aliases
CELE_B0464.5
Chromosome Location
Chr.III: 9456608 - 9463007 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025962.3 NP_001021133.1 7 - 8 1199 95
NM_001025963.6 NP_001021134.1 8 - 9 1382 95
NM_001306716.3 NP_001293645.1 4 - 5 883 95
NM_066679.5 NP_499080.3 6 - 7 2491 95
GenBank mRNA AF241656.1 - 7 - 8 1252 95

View Full Product Details