Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
com-1
Entrez Gene ID
176518
Gene Name
Completion Of Meiotic recombination (budding yeast Com) related
Gene Aliases
CELE_C44B9.5
Chromosome Location
Chr.III: 10896363 - 10901836 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066997.4 NP_499398.3 3 - 4 771 139

View Full Product Details