Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tba-7
Entrez Gene ID
176568
Gene Name
Tubulin alpha chain
Gene Aliases
CELE_T28D6.2
Chromosome Location
Chr.III: 11368351 - 11371788 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067062.7 NP_499463.1 3 - 4 699 69

View Full Product Details