Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-31
Entrez Gene ID
176572
Gene Name
protein-coding
Gene Aliases
CELE_Y47D3B.5
Chromosome Location
Chr.III: 11410678 - 11424661 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372661.1 NP_001359517.1 20 - 21 3237 93

View Full Product Details