Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-39
Entrez Gene ID
182864
Gene Name
protein-coding
Gene Aliases
CELE_C24H11.8
Chromosome Location
Chr.III: 11795748 - 11803243 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083169.2 NP_001076638.1 9 - 10 1274 88
NM_001083170.3 NP_001076639.1 9 - 10 1226 88

View Full Product Details