Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-16
Entrez Gene ID
176623
Gene Name
Nuclear Pore complex Protein
Gene Aliases
CELE_Y56A3A.17
Chromosome Location
Chr.III: 11915928 - 11917876 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067150.2 NP_499551.1 1 - 2 148 108

View Full Product Details