Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
top-3
Entrez Gene ID
176631
Gene Name
DNA topoisomerase 3
Gene Aliases
CELE_Y56A3A.27
Chromosome Location
Chr.III: 11949906 - 11960180 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067157.6 NP_499558.1 4 - 5 633 83
GenBank mRNA AF057032.1 - 4 - 5 633 83

View Full Product Details