Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snr-6
Entrez Gene ID
176668
Gene Name
Probable small nuclear ribonucleoprotein E
Gene Aliases
CELE_Y49E10.15
Chromosome Location
Chr.III: 12429155 - 12429630 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067219.3 NP_499620.1 1 - 2 72 60

View Full Product Details