Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gly-5
Entrez Gene ID
176736
Gene Name
protein-coding
Gene Aliases
CELE_Y39E4B.12
Chromosome Location
Chr.III: 13187384 - 13204903 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383020.1 8 - 9 1551 151

View Full Product Details