Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-18
Entrez Gene ID
176748
Gene Name
Nucleoporin SEH1
Gene Aliases
CELE_Y43F4B.4
Chromosome Location
Chr.III: 13296853 - 13299122 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067339.5 NP_499740.1 1 - 2 72 81

View Full Product Details