Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cua-1
Entrez Gene ID
176770
Gene Name
protein-coding
Gene Aliases
CELE_Y76A2A.2
Chromosome Location
Chr.III: 13439728 - 13457225 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268273.3 NP_001255202.1 6 - 7 867 82
NM_001268274.3 NP_001255203.1 3 - 4 368 82
GenBank mRNA D83665.1 - 5 - 6 803 82

View Full Product Details