Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqbp-1.1
Entrez Gene ID
176847
Gene Name
WW domain-containing protein
Gene Aliases
CELE_T21D12.3
Chromosome Location
Chr.IV: 259989 - 261683 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067489.5 NP_499890.2 1 - 2 221 106

View Full Product Details