Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-48
Entrez Gene ID
191980
Gene Name
Seven TM Receptor
Gene Aliases
CELE_C34D4.8
Chromosome Location
Chr.IV: 7122005 - 7124035 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068715.2 NP_501116.1 3 - 4 1119 118

View Full Product Details