Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-1
Entrez Gene ID
191967
Gene Name
Seven TM Receptor
Gene Aliases
CELE_C42D4.5
Chromosome Location
Chr.IV: 7173789 - 7175008 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068729.2 NP_501130.2 3 - 4 629 108

View Full Product Details