Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- dif-1
- Entrez Gene ID
-
177530
- Gene Name
- Protein dif-1
- Gene Aliases
-
CELE_F49E8.5
- Chromosome Location
-
Chr.IV: 7551560 - 7556422 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383120.1 |
|
8 - 9 |
4070 |
62 |
GenBank mRNA |
X76115.1 |
- |
- |
808 |
62 |
- Gene Symbol
- F49E8.6
- Entrez Gene ID
-
177531
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F49E8.6
- Chromosome Location
-
Chr.IV: 7551611 - 7556478 on Build WBcel235
View Full Product Details