Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
itr-1
Entrez Gene ID
177546
Gene Name
protein-coding
Gene Aliases
CELE_F33D4.2
Chromosome Location
Chr.IV: 7675174 - 7696930 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027999.4 NP_001023170.1 23 - 24 5850 77
NM_001028000.5 NP_001023171.1 21 - 22 5682 77
NM_001028001.5 NP_001023172.1 24 - 25 5840 77
NM_001028002.4 NP_001023173.1 22 - 23 5778 77
NM_001268376.4 NP_001255305.1 23 - 24 5775 77
GenBank mRNA AF168688.1 - 22 - 23 5778 77
AJ243179.2 - 22 - 23 5800 77
AJ243180.2 - 23 - 24 5873 77
AJ243181.2 - 22 - 23 5793 77
AJ243182.2 - 24 - 25 5860 77

View Full Product Details