Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ntp-1
Entrez Gene ID
177567
Gene Name
protein-coding
Gene Aliases
CELE_C33H5.14
Chromosome Location
Chr.IV: 7792349 - 7794809 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307237.2 NP_001294166.1 1 - 2 132 73
NM_001307238.2 NP_001294167.1 1 - 2 283 73

View Full Product Details