Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
xpo-3
Entrez Gene ID
177581
Gene Name
protein-coding
Gene Aliases
CELE_C49H3.10
Chromosome Location
Chr.IV: 7916689 - 7924032 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307224.3 NP_001294153.1 1 - 2 61 86
NM_068919.8 NP_501320.2 1 - 2 61 86

View Full Product Details