Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
csr-1
Entrez Gene ID
177591
Gene Name
protein-coding
Gene Aliases
CELE_F20D12.1
Chromosome Location
Chr.IV: 7957267 - 7962157 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047473.4 NP_001040938.1 9 - 10 2432 63
NM_001047474.3 NP_001040939.1 7 - 8 1868 63

View Full Product Details