Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sph-1
Entrez Gene ID
177608
Gene Name
MARVEL domain-containing protein
Gene Aliases
CELE_F42G8.11
Chromosome Location
Chr.IV: 8132175 - 8133692 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068959.7 NP_501360.2 3 - 4 730 113

View Full Product Details