Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-49
Entrez Gene ID
177666
Gene Name
Protein lin-49
Gene Aliases
CELE_F42A9.2
Chromosome Location
Chr.IV: 8620683 - 8626162 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069074.7 NP_501475.1 5 - 6 1578 123
GenBank mRNA AF163018.1 - 5 - 6 1563 123

View Full Product Details