Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-35
Entrez Gene ID
183159
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_C33D9.4
Chromosome Location
Chr.IV: 8788449 - 8790244 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069117.1 NP_501518.1 4 - 5 690 154

View Full Product Details