Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alp-1
Entrez Gene ID
177701
Gene Name
protein-coding
Gene Aliases
CELE_T11B7.4
Chromosome Location
Chr.IV: 8852494 - 8862246 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028199.5 NP_001023370.1 9 - 10 1469 71
NM_001028200.6 NP_001023371.1 11 - 12 3426 71

View Full Product Details