The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001028199.5 | NP_001023370.1 | 5 - 6 | 592 | 72 |
NM_001028200.6 | NP_001023371.1 | 6 - 7 | 803 | 72 | |
NM_001047522.4 | NP_001040987.1 | 6 - 7 | 809 | 72 | |
NM_001380387.1 | NP_001367079.1 | 6 - 7 | 803 | 72 | |
NM_069133.5 | NP_501534.2 | 6 - 7 | 803 | 72 |