Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rad-26
Entrez Gene ID
177706
Gene Name
protein-coding
Gene Aliases
CELE_C27B7.4
Chromosome Location
Chr.IV: 8893855 - 8901391 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380389.2 NP_001368704.1 4 - 5 736 136
NM_069144.6 NP_501545.1 5 - 6 792 136

View Full Product Details