Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vps-26
Entrez Gene ID
177764
Gene Name
protein-coding
Gene Aliases
CELE_T20D3.7
Chromosome Location
Chr.IV: 9339400 - 9341802 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136399.3 NP_001129871.1 3 - 4 374 125
NM_001276834.3 NP_001263763.1 3 - 4 536 125
NM_001380407.1 NP_001368165.1 4 - 5 499 125
NM_069243.5 NP_501644.1 3 - 4 530 125

View Full Product Details