Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wrt-3
Entrez Gene ID
177781
Gene Name
protein-coding
Gene Aliases
CELE_F38E11.7
Chromosome Location
Chr.IV: 9473633 - 9481011 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307163.3 NP_001294092.1 1 - 2 76 84
NM_001383164.1 13 - 14 2333 84

Gene Symbol
cng-3
Entrez Gene ID
3565463
Gene Name
Cyclic nucleotide-binding domain-containing protein
Gene Aliases
CELE_F38E11.12
Chromosome Location
Chr.IV: 9475836 - 9481037 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028023.3 NP_001023194.1 13 - 14 2361 84

View Full Product Details