Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-1
Entrez Gene ID
183146
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_C33A12.14
Chromosome Location
Chr.IV: 9488615 - 9490369 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069275.4 NP_501676.2 2 - 3 649 68

View Full Product Details