Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nlp-35
Entrez Gene ID
183138
Gene Name
Neuropeptide-Like Protein
Gene Aliases
CELE_C33A12.2
Chromosome Location
Chr.IV: 9518363 - 9521946 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069286.7 NP_501687.1 2 - 3 215 85

View Full Product Details