Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
twk-9
Entrez Gene ID
177803
Gene Name
protein-coding
Gene Aliases
CELE_ZK1251.8
Chromosome Location
Chr.IV: 9693174 - 9697005 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069323.3 NP_501724.2 1 - 2 66 66

View Full Product Details