Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-9
Entrez Gene ID
177803
Gene Name
protein-coding
Gene Aliases
CELE_ZK1251.8
Chromosome Location
Chr.IV: 9693174 - 9697005 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307073.3 NP_001294002.1 6 - 7 930 84
NM_069323.3 NP_501724.2 6 - 7 958 84

View Full Product Details