Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-4
Entrez Gene ID
188477
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_T13F2.5
Chromosome Location
Chr.IV: 9780955 - 9784789 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069345.6 NP_501746.3 5 - 6 708 114

View Full Product Details