Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dct-15
Entrez Gene ID
189289
Gene Name
protein-coding
Gene Aliases
CELE_W08D2.3
Chromosome Location
Chr.IV: 9806406 - 9810733 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047534.5 NP_001040999.2 6 - 7 1051 119
NM_001268500.2 NP_001255429.1 6 - 7 1068 119

View Full Product Details