Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glt-3
Entrez Gene ID
177882
Gene Name
Putative sodium-dependent excitatory amino acid transporter glt-3
Gene Aliases
CELE_K08F4.4
Chromosome Location
Chr.IV: 10130257 - 10133283 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069443.6 NP_501844.1 4 - 5 706 154

View Full Product Details