Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
egl-21
Entrez Gene ID
177940
Gene Name
Carboxypeptidase E
Gene Aliases
CELE_F01D4.4
Chromosome Location
Chr.IV: 10478442 - 10481844 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069534.7 NP_501935.1 1 - 2 259 143

View Full Product Details