Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-21
Entrez Gene ID
177940
Gene Name
Carboxypeptidase E
Gene Aliases
CELE_F01D4.4
Chromosome Location
Chr.IV: 10478442 - 10481844 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069534.7 NP_501935.1 4 - 5 1439 53

View Full Product Details