Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-24
Entrez Gene ID
176898
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_Y55F3C.8
Chromosome Location
Chr.IV: 728500 - 731037 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067575.3 NP_499976.3 5 - 6 878 128

View Full Product Details