Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
immp-2
Entrez Gene ID
176919
Gene Name
protein-coding
Gene Aliases
CELE_Y55F3AM.8
Chromosome Location
Chr.IV: 1003595 - 1005861 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380028.1 NP_001368250.1 1 - 2 111 73
NM_067621.4 NP_500022.1 2 - 3 218 73

View Full Product Details