Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trap-3
Entrez Gene ID
177028
Gene Name
TRanslocon-Associated Protein
Gene Aliases
CELE_Y38F2AR.2
Chromosome Location
Chr.IV: 2389445 - 2393032 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067797.7 NP_500198.1 1 - 2 160 67

View Full Product Details