Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdh-10
Entrez Gene ID
183476
Gene Name
CaDHerin family
Gene Aliases
CELE_C45G7.5
Chromosome Location
Chr.IV: 2436498 - 2463369 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067803.6 NP_500204.3 17 - 18 5276 89

View Full Product Details