Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sprr-3
Entrez Gene ID
190541
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y69A2AR.15
Chromosome Location
Chr.IV: 2542301 - 2549952 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067817.4 NP_500218.2 10 - 11 1121 113

View Full Product Details