Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ngn-1
Entrez Gene ID
177045
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_Y69A2AR.29
Chromosome Location
Chr.IV: 2626062 - 2628271 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067835.5 NP_500236.1 1 - 2 151 111

View Full Product Details