Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
otpl-1
Entrez Gene ID
181841
Gene Name
protein-coding
Gene Aliases
CELE_B0212.1
Chromosome Location
Chr.IV: 3568919 - 3577770 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307511.1 NP_001294440.1 9 - 10 1202 88
NM_001380150.1 NP_001368270.1 10 - 11 1262 88
NM_067973.4 NP_500374.2 5 - 6 814 88

View Full Product Details