Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-45
Entrez Gene ID
177139
Gene Name
protein-coding
Gene Aliases
CELE_Y37E11AR.5
Chromosome Location
Chr.IV: 3748210 - 3751101 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307366.3 NP_001294295.1 2 - 3 135 81
NM_068009.7 NP_500410.2 2 - 3 231 81

View Full Product Details