Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-21
Entrez Gene ID
185417
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F37C4.1
Chromosome Location
Chr.IV: 3889263 - 3892563 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068032.3 NP_500433.3 11 - 12 1788 139

View Full Product Details