Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- clec-75
- Entrez Gene ID
-
189949
- Gene Name
- C-type lectin domain-containing protein
- Gene Aliases
-
CELE_Y46C8AL.9
- Chromosome Location
-
Chr.IV: 3960734 - 3964204 on Build WBcel235
- Gene Symbol
- Y46C8AM.1
- Entrez Gene ID
-
34700653
- Gene Name
- C-type lectin domain-containing protein
- Gene Aliases
-
CELE_Y46C8AM.1
- Chromosome Location
-
Chr.IV: 3960732 - 3964384 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383071.1 |
|
2 - 3 |
502 |
112 |
View Full Product Details