Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hlh-30
Entrez Gene ID
177157
Gene Name
protein-coding
Gene Aliases
CELE_W02C12.3
Chromosome Location
Chr.IV: 4014787 - 4034940 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028240.6 NP_001023411.1 9 - 10 1367 89
NM_001028242.6 NP_001023413.1 7 - 8 1071 89
NM_001028243.3 NP_001023414.1 7 - 8 1236 89
NM_001364041.3 NP_001350991.1 8 - 9 1098 89
NM_001380173.1 NP_001368701.1 9 - 10 1173 89
NM_001383072.1 9 - 10 1336 89
NM_001383073.1 9 - 10 1307 89
NM_068060.5 NP_500461.1 8 - 9 1292 89
NM_068061.6 NP_500462.1 8 - 9 1309 89

View Full Product Details