Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hlh-30
Entrez Gene ID
177157
Gene Name
protein-coding
Gene Aliases
CELE_W02C12.3
Chromosome Location
Chr.IV: 4014787 - 4034940 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028240.6 NP_001023411.1 3 - 4 503 63
NM_001372787.3 NP_001360026.1 2 - 3 447 63
NM_001380173.1 NP_001368701.1 3 - 4 309 63
NM_001383072.1 3 - 4 472 63
NM_001383073.1 3 - 4 443 63
NM_068061.6 NP_500462.1 2 - 3 445 63

View Full Product Details