Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-55
Entrez Gene ID
189151
Gene Name
F-box B protein
Gene Aliases
CELE_W03D2.2
Chromosome Location
Chr.IV: 4068780 - 4069548 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068072.1 NP_500473.1 1 - 2 184 125

View Full Product Details